Learn More
Aviva Systems Biology Corp IDH3A Antibody - middle region (ARP42238_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42238_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IDH3A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human IDH3A. Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast
Catalog No. 50-014-58159