Learn More
Enzo Life Sciences IDO (human) polyclonal antibody (100µg)
SDP
Catalog No. 502007100
Encompass_Preferred
Host: Sheep. Immunogen: Recombinant human IDO. Species reactivity: Human. Applications: ELISA, Flow Cytometry, IHC, WB. Formulation: Liquid. 0.2 µm filtered antibody solution in PBS, containing 0.02% sodium azide and 0.1% bovine serum albumin.
Catalog No. 50-200-7100 Supplier Enzo Life Sciences Supplier No. ADI9083080100
Host: Sheep. Immunogen: Recombinant human IDO. Species reactivity: Human. Applications: ELISA, Flow Cytometry, IHC, WB. Formulation: Liquid. 0.2 µm filtered antibody solution in PBS, containing 0.02% sodium azide and 0.1% bovine serum albumin.