Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC IDO-IN-12 100mg | 1888341-29-4 | 100 MG
SDP

Catalog No. 5000226885
Encompass_Preferred

IDO-IN-12 is a small-molecule inhibitor of indoleamine 2,3-dioxygenase (IDO) disclosed in patent WO 2017181849 A1. It is supplied as a white to off-white solid for preclinical IDO pathway research and biochemical assays.

  • Small-molecule IDO inhibitor for preclinical research.
  • CAS number 1888341-29-4.
  • Molecular formula C13H11BrFN5O3S.
  • Molecular weight 416.23.
  • Appearance white to off-white solid.
  • Purity 99.22% (reported).
  • Available in multiple sizes including 100 MG and solution formats.
  • Storage: powder -20°C (up to 3 years) or 4°C (up to 2 years); in solvent -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-26885 Supplier Medchemexpress LLC Supplier No. HY115122100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

IDO-IN-12 is a small-molecule inhibitor of indoleamine 2,3-dioxygenase (IDO) disclosed in patent WO 2017181849 A1. It is supplied as a white to off-white solid for preclinical IDO pathway research and biochemical assays.

  • Small-molecule IDO inhibitor for preclinical research.
  • CAS number 1888341-29-4.
  • Molecular formula C13H11BrFN5O3S.
  • Molecular weight 416.23.
  • Appearance white to off-white solid.
  • Purity 99.22% (reported).
  • Available in multiple sizes including 100 MG and solution formats.
  • Storage: powder -20°C (up to 3 years) or 4°C (up to 2 years); in solvent -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.