Learn More
Medchemexpress LLC HY-18769 5mg Medchemexpress, IDO-IN-4 CAS:1629125-65-0 Purity:>98%
SDP
Catalog No. 502082086
Encompass_Preferred
Medchemexpress, HY-18769 5mg IDO-IN-4 CAS:1629125-65-0 IDO-IN-4 is an indoleamine 2,3-dioxygenase 1 (IDO-1) inhibitor, extracted from patent WO2014150677A1, Compound example 1 enantiomer 1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2086 Supplier Medchemexpress LLC Supplier No. HY187695MG
Medchemexpress, HY-18769 5mg IDO-IN-4 CAS:1629125-65-0 IDO-IN-4 is an indoleamine 2,3-dioxygenase 1 (IDO-1) inhibitor, extracted from patent WO2014150677A1, Compound example 1 enantiomer 1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.