Learn More
Medchemexpress LLC HY-18770A 5mg Medchemexpress, IDO-IN-6 CAS:1402837-76-6 Purity:>98%
SDP
Catalog No. 501932821
Encompass_Preferred
Medchemexpress, HY-18770A 5mg IDO-IN-6 CAS:1402837-76-6 IDO-IN-6 (NLG-1486) is an indoleamine 2,3-dioxygenase ( IDO ) inhibitor extracted from patent WO WO2012142237A1, Compound 1486, has an IC 50 of <1 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2821 Supplier Medchemexpress LLC Supplier No. HY18770A5MG
Medchemexpress, HY-18770A 5mg IDO-IN-6 CAS:1402837-76-6 IDO-IN-6 (NLG-1486) is an indoleamine 2,3-dioxygenase ( IDO ) inhibitor extracted from patent WO WO2012142237A1, Compound 1486, has an IC 50 of <1 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.