Learn More
Medchemexpress LLC HY-18770C 5mg Medchemexpress, IDO-IN-8 CAS:1402837-77-7 Purity:>98%
SDP
Catalog No. 501932822
Encompass_Preferred
Medchemexpress, HY-18770C 5mg IDO-IN-8 CAS:1402837-77-7 IDO-IN-8 (NLG-1487) is an indoleamine 2,3-dioxygenase ( IDO ) inhibitor extracted from patent WO WO2012142237A1, compound 1487, has an IC 50 of 1-10 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2822 Supplier Medchemexpress LLC Supplier No. HY18770C5MG
Medchemexpress, HY-18770C 5mg IDO-IN-8 CAS:1402837-77-7 IDO-IN-8 (NLG-1487) is an indoleamine 2,3-dioxygenase ( IDO ) inhibitor extracted from patent WO WO2012142237A1, compound 1487, has an IC 50 of 1-10 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.