Learn More
Medchemexpress LLC HY-111540 10mg Medchemexpress, IDO1-IN-5 CAS:2166616-75-5 Purity:>98%
SDP
Catalog No. 501968905
Encompass_Preferred
Medchemexpress, HY-111540 10mg IDO1-IN-5 CAS:2166616-75-5 IDO1-IN-5 is a potent, selective and brain penetrated inhibitor of IDO1 activity, binds to apo-IDO1 lacking heme rather than mature heme-bound IDO1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8905 Supplier Medchemexpress LLC Supplier No. HY11154010MG
Medchemexpress, HY-111540 10mg IDO1-IN-5 CAS:2166616-75-5 IDO1-IN-5 is a potent, selective and brain penetrated inhibitor of IDO1 activity, binds to apo-IDO1 lacking heme rather than mature heme-bound IDO1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.