Learn More
Medchemexpress LLC Ido1/tdo-in-4 | 461424-21-5 | C14H10N4 | 5 MG
SDP
Catalog No. 5000375886
Encompass_Preferred
IDO1/TDO-IN-4 is a potent IDO1/TDO dual inhibitor, with IC50 values of 3.53 μM (IDO1) and 1.15 μM (TDO). It forms a hydrogen bond with IDO1 and π-π stacking interaction with TDO. This product can be used in the research of depression, and depression-induced infectious, metabolic, and autoimmune disorders.
  • Potent IDO1/TDO dual inhibitor.
  • IC50 values: 3.53 μM (IDO1) and 1.15 μM (TDO).
  • Forms hydrogen bond with IDO1 and π-π stacking interaction with TDO.
  • Can be used in the research of depression, and depression-induced infectious, metabolic, and autoimmune disorders.
Catalog No. 50-003-75886 Supplier Medchemexpress LLC Supplier No. HY1511085MG
IDO1/TDO-IN-4 is a potent IDO1/TDO dual inhibitor, with IC50 values of 3.53 μM (IDO1) and 1.15 μM (TDO). It forms a hydrogen bond with IDO1 and π-π stacking interaction with TDO. This product can be used in the research of depression, and depression-induced infectious, metabolic, and autoimmune disorders.
  • Potent IDO1/TDO dual inhibitor.
  • IC50 values: 3.53 μM (IDO1) and 1.15 μM (TDO).
  • Forms hydrogen bond with IDO1 and π-π stacking interaction with TDO.
  • Can be used in the research of depression, and depression-induced infectious, metabolic, and autoimmune disorders.