Learn More
Abcam Ido x uridine, Antiviral thymidine analog, 1G
Catalog No. 502706217
Encompass_Preferred
MW 354.1 Da, Purity >98%. Selective cytotoxic cell-permeable anti-herpesvirus agent (IC₅₀ = 4.3 μM). Radiosensitizing. Incorporates into DNA increases susceptibility of cancer cell to radiation damage. Active in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6217 Supplier Abcam Supplier No. AB1425811G
MW 354.1 Da, Purity >98%. Selective cytotoxic cell-permeable anti-herpesvirus agent (IC₅₀ = 4.3 μM). Radiosensitizing. Incorporates into DNA increases susceptibility of cancer cell to radiation damage. Active in vivo. .
The product is subject to the following: Abcam Restricted Use Statement