Learn More
Cayman Chemical IDR 1002trIfluoROCTAte sa 10mg
SDP
Catalog No. 502914943
Encompass_Preferred
A synthetic cationic innate defense regulator peptide; increases the production of CCL2, CCL7, and CXCL1 in isolated human PBMCs at 100 UG/ml; increases the numbers of neutrophils and monocytes in mouse peritoneum and protects against S. aureus and E. coli infections in mice at 200 UG/animal
Catalog No. 50-291-4943 Supplier Cayman Chemical Supplier No. 3689310 MG
A synthetic cationic innate defense regulator peptide; increases the production of CCL2, CCL7, and CXCL1 in isolated human PBMCs at 100 UG/ml; increases the numbers of neutrophils and monocytes in mouse peritoneum and protects against S. aureus and E. coli infections in mice at 200 UG/animal