Learn More
Cayman Chemical IDR 1002 (trifluoroacetate salt), Purity: ≥95%, 50 mg
SDP
Catalog No. 502914945
Encompass_Preferred
A synthetic cationic innate defense regulator peptide; increases the production of CCL2, CCL7, and CXCL1 in isolated human PBMCs at 100 µg/ml; increases the numbers of neutrophils and monocytes in mouse peritoneum and protects against S. aureus and E. coli infections in mice at 200 µg/animal
Catalog No. 50-291-4945 Supplier Cayman Chemical Supplier No. 3689350 MG
A synthetic cationic innate defense regulator peptide; increases the production of CCL2, CCL7, and CXCL1 in isolated human PBMCs at 100 µg/ml; increases the numbers of neutrophils and monocytes in mouse peritoneum and protects against S. aureus and E. coli infections in mice at 200 µg/animal