Learn More
Medchemexpress LLC IDR-1018 acetate | 99.6% | 1535.93 (free base) | 10 MG
SDP
Catalog No. 5000398695
Encompass_Preferred
IDR-1018 acetate is an innate defense regulator conjugate with MICs of 16 μg/mL for MRSA USA300 LAC, MRSA SAP 0017, and S. epidermidis ATCC14990.
  • Can be used to synthesize V-IDR1018 (vancomycin-innate defense regulator conjugate).
  • Modulates the expression of neutrophil adhesion and activation markers in vitro.
  • Potent inducer of chemokines.
  • Demonstrates anti-infective and anti-inflammatory activity in mouse models.
  • Effective in treating Plasmodium berghei ANKA cerebral malaria when combined with standard first-line antimalarials.
Catalog No. 50-003-98695 Supplier Medchemexpress LLC Supplier No. HYP3361A10MG
IDR-1018 acetate is an innate defense regulator conjugate with MICs of 16 μg/mL for MRSA USA300 LAC, MRSA SAP 0017, and S. epidermidis ATCC14990.
  • Can be used to synthesize V-IDR1018 (vancomycin-innate defense regulator conjugate).
  • Modulates the expression of neutrophil adhesion and activation markers in vitro.
  • Potent inducer of chemokines.
  • Demonstrates anti-infective and anti-inflammatory activity in mouse models.
  • Effective in treating Plasmodium berghei ANKA cerebral malaria when combined with standard first-line antimalarials.