Learn More
Medchemexpress LLC Idramantone | 20098-14-0 | 99.94% | 200 MG
SDP
Catalog No. 5000413855
Encompass_Preferred
Idramantone is an Adamantane derivative and an immunostimulant. It serves as a versatile starting material for synthesizing various adamantine derivatives. It can be produced using P450cam monooxygenase coupled with NADH regeneration as an oxidation biocatalyst and can be studied in research on bronchial pathology.
  • Adamantane derivative
  • Immunostimulant
  • Versatile starting material for synthesizing various derivatives
  • Can be produced using P450cam monooxygenase
  • Suitable for research on bronchial pathology
Catalog No. 50-004-13855 Supplier Medchemexpress LLC Supplier No. HYB1044200MG
Idramantone is an Adamantane derivative and an immunostimulant. It serves as a versatile starting material for synthesizing various adamantine derivatives. It can be produced using P450cam monooxygenase coupled with NADH regeneration as an oxidation biocatalyst and can be studied in research on bronchial pathology.
  • Adamantane derivative
  • Immunostimulant
  • Versatile starting material for synthesizing various derivatives
  • Can be produced using P450cam monooxygenase
  • Suitable for research on bronchial pathology