Learn More
Apexbio Technology LLC IDRAMANTONE-200MG
SDP
Catalog No. 5000570652
Encompass_Preferred
Idramantone (CAS No 20098-14-0) is an intriguing compound in the field of biomedical research known for its potential applications in studying cellular signaling pathways Derived from a synthetic origin Idramantone primarily targets the modulation of key molecular pathways involved in cellular metabolism and stress responses Its mechanism of action is linked to the inhibition of certain enzymes and receptors influencing cell proliferation and differentiation In vitro studies demonstrate that Idramantone exhibits activity in the nanomolar to low micromolar range making it significant for precise biochemical analyses This compound is commonly employed in cell models and animal studies serving as a valuable tool in drug screening and molecular biology research Experimental concentrations typically depend on the specific research design allowing flexibility in application to suit various investigative needs
Catalog No. 50-005-70652 Supplier Apexbio Technology LLC Supplier No. C6638200
Idramantone (CAS No 20098-14-0) is an intriguing compound in the field of biomedical research known for its potential applications in studying cellular signaling pathways Derived from a synthetic origin Idramantone primarily targets the modulation of key molecular pathways involved in cellular metabolism and stress responses Its mechanism of action is linked to the inhibition of certain enzymes and receptors influencing cell proliferation and differentiation In vitro studies demonstrate that Idramantone exhibits activity in the nanomolar to low micromolar range making it significant for precise biochemical analyses This compound is commonly employed in cell models and animal studies serving as a valuable tool in drug screening and molecular biology research Experimental concentrations typically depend on the specific research design allowing flexibility in application to suit various investigative needs