Learn More
Aviva Systems Biology Corp IDS Antibody - N-terminal region (ARP60956_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60956_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IDS. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human IDS. Alternate names: MPS2, SIDSApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-287-211