Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Iduronate 2-sulfatase | 50936-59-9 | >95.0% | 500 UG
SDP

Catalog No. 5000290070
Encompass_Preferred

Recombinant human iduronate 2-sulfatase (IDS) is a lysosomal enzyme produced in HEK293 cells and supplied lyophilized as an active, His-tagged protein for biochemical and enzymatic studies. It hydrolyzes the C2-sulfate ester of glycosaminoglycans and is provided with validated purity and activity specifications.

  • Expressed in HEK293 cells.
  • C-terminal 10xHis tag for purification.
  • Purity >95.0% by reducing SDS-PAGE.
  • Specific activity 6.123 pmoL/min/μg (PNCS substrate assay).
  • Lyophilized from 0.22 μm filtered PBS, pH 7.4.
  • Approximate molecular weight 75-95 kDa due to glycosylation.
  • Store at -20°C; after reconstitution stable at 4°C for 1 week.
  • Available in a 500 μg size.

Catalog No. 50-002-90070 Supplier Medchemexpress LLC Supplier No. HYP76399500UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human iduronate 2-sulfatase (IDS) is a lysosomal enzyme produced in HEK293 cells and supplied lyophilized as an active, His-tagged protein for biochemical and enzymatic studies. It hydrolyzes the C2-sulfate ester of glycosaminoglycans and is provided with validated purity and activity specifications.

  • Expressed in HEK293 cells.
  • C-terminal 10xHis tag for purification.
  • Purity >95.0% by reducing SDS-PAGE.
  • Specific activity 6.123 pmoL/min/μg (PNCS substrate assay).
  • Lyophilized from 0.22 μm filtered PBS, pH 7.4.
  • Approximate molecular weight 75-95 kDa due to glycosylation.
  • Store at -20°C; after reconstitution stable at 4°C for 1 week.
  • Available in a 500 μg size.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.