Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Iduronate 2-sulfatase protein, human | 50936-59-9 | >95.0% | 1 MG
SDP

Catalog No. 5000290213
Encompass_Preferred

Recombinant human iduronate 2-sulfatase (IDS) is a lysosomal enzyme produced in HEK293 cells and supplied lyophilized for research use. It participates in the degradation of dermatan sulfate and heparan sulfate, and is suitable for biochemical assays and enzyme activity studies.

  • Expression system: HEK293 cells
  • Tag: C-terminal His
  • Purity: >95.0% by reducing SDS-PAGE
  • Molecular weight: ~75-95 kDa (glycosylated)
  • Formulation: lyophilized from 0.22 μm filtered PBS, pH 7.4
  • Storage: store lyophilized at -20°C; aliquot and freeze at -80°C for long term
  • Activity: specific activity ~6.123 pmoL/min/μg

Catalog No. 50-002-90213 Supplier Medchemexpress LLC Supplier No. HYP763991MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human iduronate 2-sulfatase (IDS) is a lysosomal enzyme produced in HEK293 cells and supplied lyophilized for research use. It participates in the degradation of dermatan sulfate and heparan sulfate, and is suitable for biochemical assays and enzyme activity studies.

  • Expression system: HEK293 cells
  • Tag: C-terminal His
  • Purity: >95.0% by reducing SDS-PAGE
  • Molecular weight: ~75-95 kDa (glycosylated)
  • Formulation: lyophilized from 0.22 μm filtered PBS, pH 7.4
  • Storage: store lyophilized at -20°C; aliquot and freeze at -80°C for long term
  • Activity: specific activity ~6.123 pmoL/min/μg

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.