Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Iduronate 2-sulfatase protein, human (HEK293, His) | >95.0% | 50 UG
SDP

Catalog No. 5000290422
Encompass_Preferred

Recombinant human iduronate 2-sulfatase (IDS) expressed in HEK293 cells and supplied as a C-terminal His-tagged lyophilized protein for research use. The enzyme catalyzes degradation of dermatan sulfate and heparan sulfate and is provided with high purity and defined activity for biochemical assays and protein characterization.

  • Supplied as lyophilized powder for convenient storage and transport.
  • Expressed in HEK293 for human-like post-translational modifications.
  • C-terminal His tag enables affinity purification and detection.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Specific activity reported as 6.123 pmoL/min/μg for functional assays.
  • Low endotoxin level (<1 EU/μg) suitable for sensitive applications.
  • Stable at -20°C; reconstitute above 100 μg/mL and add carrier protein for long-term storage.

Catalog No. 50-002-90422 Supplier Medchemexpress LLC Supplier No. HYP7639950UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human iduronate 2-sulfatase (IDS) expressed in HEK293 cells and supplied as a C-terminal His-tagged lyophilized protein for research use. The enzyme catalyzes degradation of dermatan sulfate and heparan sulfate and is provided with high purity and defined activity for biochemical assays and protein characterization.

  • Supplied as lyophilized powder for convenient storage and transport.
  • Expressed in HEK293 for human-like post-translational modifications.
  • C-terminal His tag enables affinity purification and detection.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Specific activity reported as 6.123 pmoL/min/μg for functional assays.
  • Low endotoxin level (<1 EU/μg) suitable for sensitive applications.
  • Stable at -20°C; reconstitute above 100 μg/mL and add carrier protein for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.