Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Iduronate 2-sulfatase (recombinant human) | 50936-59-9 | >95.0% | 100 UG
SDP

Catalog No. 5000290486
Encompass_Preferred

Recombinant human iduronate 2-sulfatase (IDS) expressed in HEK293 cells and supplied as a His-tagged, lyophilized protein for research use. IDS catalyzes removal of 2-sulfate groups from iduronate residues in glycosaminoglycans; the product includes documented purity, endotoxin, activity, and storage specifications to support biochemical and cell-based assays.

  • Expressed in HEK293 cells for human-like glycosylation
  • C-terminal His tag enables detection and purification
  • Lyophilized from PBS (pH 7.4) for stability and convenient handling
  • Purity greater than 95% by reducing SDS-PAGE
  • Endotoxin level <1 EU/μg, suitable for many cell-based assays
  • Specific activity 6.123 pmoL/min/μg measured with PNCS substrate
  • Store at -20°C; aliquot and keep at -80°C for extended storage

Catalog No. 50-002-90486 Supplier Medchemexpress LLC Supplier No. HYP76399100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human iduronate 2-sulfatase (IDS) expressed in HEK293 cells and supplied as a His-tagged, lyophilized protein for research use. IDS catalyzes removal of 2-sulfate groups from iduronate residues in glycosaminoglycans; the product includes documented purity, endotoxin, activity, and storage specifications to support biochemical and cell-based assays.

  • Expressed in HEK293 cells for human-like glycosylation
  • C-terminal His tag enables detection and purification
  • Lyophilized from PBS (pH 7.4) for stability and convenient handling
  • Purity greater than 95% by reducing SDS-PAGE
  • Endotoxin level <1 EU/μg, suitable for many cell-based assays
  • Specific activity 6.123 pmoL/min/μg measured with PNCS substrate
  • Store at -20°C; aliquot and keep at -80°C for extended storage

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.