Learn More
TARGETMOL CHEMICALS INC IDX184 5MG
SDP
Catalog No. 502580600
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg and bulk. Please contact Fisher for quotes. IDX184 is a potent orally active targeted HCV polymerase inhibitor and nucleoside polymerase.IDX184 effectively inhibits HCV polymerase (IC50=0.31 uM Ki=52.3 nM). IDX184 is a liver-targeted nucleotide prodrug with anti-HCV activity that inhibits HCV NS5B polymerase and can be used in combination with pegylated interferon-(alpha)2a and ribavirin for the primary treatment of chronic hepatitis C. It has been shown to inhibit HCV polymerase and nucleoside polymerase. purity: 99%
Catalog No. 50-258-0600 Supplier TARGETMOL CHEMICALS INC Supplier No. T275865MG
Also available in 1 mg 10 mg 25 mg 50 mg and bulk. Please contact Fisher for quotes. IDX184 is a potent orally active targeted HCV polymerase inhibitor and nucleoside polymerase.IDX184 effectively inhibits HCV polymerase (IC50=0.31 uM Ki=52.3 nM). IDX184 is a liver-targeted nucleotide prodrug with anti-HCV activity that inhibits HCV NS5B polymerase and can be used in combination with pegylated interferon-(alpha)2a and ribavirin for the primary treatment of chronic hepatitis C. It has been shown to inhibit HCV polymerase and nucleoside polymerase. purity: 99%