Learn More
Thermo Scientific Viper Capillary MP35N, 0.100 mm x 250 mm
Supplier:  Thermo Scientific 6042.2330
Encompass
Viper Capillary MP35N, 0.100 mm x 250 mm, for use with all HPLC and UHPLC Systems, 1500 bar maximum pressure
Catalog No. NC0917985