Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ IDXpert™ Hard Cases

Catalog No. 19104463 Shop All Brady Corporation Products
Change view
Click to view available options
For use with keyboard layout
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-104-463 For use with keyboard layout
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-104-463 Supplier Brady™ Supplier No. XPERTHCKEY

missing translation for 'validMainframeMsg'

IDXpert™ Hard Cases are for use with ABC and Keyboard layout.

  • Package includes the Case only (printer or keyboard sold separately)
  • Designed for use with the BMP™41 Label Printer from Brady™
  • The durable hard case protects your printer or keyboard against impacts and dust while allowing for easy mobility
  • Keeps printer accessories organized and on hand

Specifications

Color Black
For Use With (Equipment) Keyboard Layout
Product Type Hard Case
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.