Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp IEEE-488 Cable, 3.3ft. (1m)

Catalog No. 50793763 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

IEEE-488 Cable, 3.3ft. (1m)

Catalog No. 50-793-763 Supplier Waters Corp Supplier No. WAT087198
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

IEEE-488 Cable, 3.3ft. (1m)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.