Learn More
Bioworld IEF Cathode Buffer (10X), 250 mL
Catalog No. 50196655
Encompass_Preferred
Isoelectrofocusing Cathode Buffer (10X) bioWORLD running buffers are prepared using high quality reagents so you can be asured of accurate test results Technical Information:Dilute with deionized water to make 1X.1X Concentration Contains: 20mM Arginine (free base) and 20mM Lysine, (free base).pH 10.1
Catalog No. 50-196-655 Supplier Bioworld Supplier No. 105300041
Isoelectrofocusing Cathode Buffer (10X) bioWORLD running buffers are prepared using high quality reagents so you can be asured of accurate test results Technical Information:Dilute with deionized water to make 1X.1X Concentration Contains: 20mM Arginine (free base) and 20mM Lysine, (free base).pH 10.1