Learn More
Crescent Chemical Co Inc IEF SAMPLE BUFFER (2X) STERILE
SDP
Catalog No. 50248600
Encompass_Preferred
IEF sample buffer (2x) sterile filtered SERVA IEF sample buffer is suited to all vertical and horizontal IEF applications and systems. The sample buffer is supplied as 2x concentrate. It is sterile filtered, beneficial to long shelf life and absence of contaminants. Sim
Catalog No. 50-248-600 Supplier Crescent Chemical Co Inc Supplier No. SE42537.01
IEF sample buffer (2x) sterile filtered SERVA IEF sample buffer is suited to all vertical and horizontal IEF applications and systems. The sample buffer is supplied as 2x concentrate. It is sterile filtered, beneficial to long shelf life and absence of contaminants. Sim