Learn More
Bioworld IEF Sample Buffer (4X), 50 mL
Catalog No. 50196658
Encompass_Preferred
Isoelectrofocusing Sample Buffer (4X) Sample buffer used in protein analysis protocols Technical Information:4X Concentration Contains: 20 mM Lysine, free base, 15% Glycerol
Catalog No. 50-196-658 Supplier Bioworld Supplier No. 105300052
Isoelectrofocusing Sample Buffer (4X) Sample buffer used in protein analysis protocols Technical Information:4X Concentration Contains: 20 mM Lysine, free base, 15% Glycerol