Learn More
Bioworld IEF Sample Buffer (4X), 25 mL
Catalog No. 50196657
Encompass_Preferred
Isoelectrofocusing Sample Buffer (4X) Sample buffer used in protein analysis protocols Technical Information:4X Concentration Contains: 20 mM Lysine, free base, 15% Glycerol
Catalog No. 50-196-657 Supplier Bioworld Supplier No. 105300051
Isoelectrofocusing Sample Buffer (4X) Sample buffer used in protein analysis protocols Technical Information:4X Concentration Contains: 20 mM Lysine, free base, 15% Glycerol