Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich IEICO-4CL ≥ 99 %

Catalog No. 913774100MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
91-377-4100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 91-377-4100MG Supplier Sigma Aldrich Supplier No. 913774100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

IEICO-4Cl is a transparent, low-bandgap non-fullerene acceptor. It is a chlorinated derivative of IEICO, which imparts it with a lower optical bandgap and lower energy levels. The weak photon absorption in the visible spectrum of IEICO-4Cl, allows high-performance organic solar cells to be fabricated with outstanding transparency. Furthermore, changing the polymer acceptor pairing with IEICO-4Cl allows tuning of the color of the solar cell device fabricated.

Specifications

CAS 2240998-88-1
Molecular Formula C114H114Cl4N4O4S4
Linear Formula C114H114Cl4N4O4S4
Quantity 100 mg
Synonym 2,2 -((2 Z,2 Z)-(((4,4,9-tris(4-hexylphenyl)-9-(4-pentylphenyl)-4,9-dihydro-s-indaceno[1,2-b:5,6-b dithiophene-2,7-diyl)bis(4-((2-ethylhexyl)oxy)thiophene 5,2-diyl))bis(methanylylidene))bis(5,6-dichloro-3-oxo-2,3-dihydro-1 H-indene-2,1-diylidene))dimalononitrile
Molecular Weight (g/mol) 1874.22
Percent Purity ≥99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.