Learn More
Selleck Chemical LLC IEM 1754 2HBr S2860-10mg
SDP
Catalog No. 503109622
Encompass_Preferred
IEM 1754 2HBr is a selective AMPA/kainate receptor blockers for GluR1 and GluR3 with IC50 of 6 M
Catalog No. 50-310-9622 Supplier Selleck Chemical LLC Supplier No. S286010MG
IEM 1754 2HBr is a selective AMPA/kainate receptor blockers for GluR1 and GluR3 with IC50 of 6 M