Learn More
Medchemexpress LLC Iem-1925 bromide | 258282-23-4 | MFCD19690950 | 98.0% | 422.24 g/mol | C17H30Br2N2 | 10 MG
SDP
Catalog No. 5000247542
Encompass_Preferred
IEM-1925 bromide is the bromide salt of a small-molecule glutamate receptor antagonist provided for research use. It is supplied as a high-purity solid suitable for in vitro and in vivo studies, with a molecular weight of 422.24 g/mol and typical purity of 98.0%. Intended for laboratory research and not for human use.
  • Glutamate receptor antagonist activity
  • High purity solid powder for reliable results
  • Suitable for in vitro and in vivo research applications
  • Available in small milligram package sizes for early-stage studies
  • Store sealed at 4°C; in solution store at -80°C for long-term
Catalog No. 50-002-47542 Supplier Medchemexpress LLC Supplier No. HY10323210MG
IEM-1925 bromide is the bromide salt of a small-molecule glutamate receptor antagonist provided for research use. It is supplied as a high-purity solid suitable for in vitro and in vivo studies, with a molecular weight of 422.24 g/mol and typical purity of 98.0%. Intended for laboratory research and not for human use.
  • Glutamate receptor antagonist activity
  • High purity solid powder for reliable results
  • Suitable for in vitro and in vivo research applications
  • Available in small milligram package sizes for early-stage studies
  • Store sealed at 4°C; in solution store at -80°C for long-term