Learn More
Aviva Systems Biology Corp IER2 Antibody - N-terminal region (ARP34401_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34401_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IER2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human IER2. Alternate names: ETR101, MGC111472, MGC15265ApplicationIHC, WB. Species reactivity: Human, Yeast
Catalog No. 50-014-55380