Learn More
Aviva Systems Biology Corp Ier3ip1 Antibody - N-terminal region (ARP47215_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47215_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Ier3ip1. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 1110057H19Rik, AI644142, AL022842, AL022863, AV026606ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-290