Learn More
Aviva Systems Biology Corp Ier5l Antibody - C-terminal region (ARP55969_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55969_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Ier5l. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 2610524G09RikApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rat
Catalog No. 50-286-101