Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp WATERS CORPORATION IEX Anion Exchange Standard, 3 Proteins, Column Benchmarking, Hi Res IEX

Catalog No. NC1781047 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This IEX Anion Exchange Standard is a specific mix containing three proteins, designed for benchmarking anion exchange column performance. It is recommended for regular use to monitor column and system performance, especially with high-resolution IEX columns, and can be used for proteins that resolve well on anion-exchange columns.

  • Contains three proteins for benchmarking anion exchange columns.
  • Designed for use with high-resolution IEX columns for monitoring performance.
  • Utilizes ion exchange chromatography with UV detection.
  • Solid formulation, supplied lyophilized in 20 mM Tris Buffer, pH 8–9.
  • Reconstitute with 1 mL of 100% Milli-Q Water.
  • Includes Horse Myoglobin (0.5 mg/mL), Conalbumin (1.25 mg/mL), and Soybean Trypsin Inhibitor (2 mg/mL).
  • Shelf life of 12 months; ships ambient.

Catalog No. NC1781047 Supplier Waters Corp Supplier No. 186006869
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This IEX Anion Exchange Standard is a specific mix containing three proteins, designed for benchmarking anion exchange column performance. It is recommended for regular use to monitor column and system performance, especially with high-resolution IEX columns, and can be used for proteins that resolve well on anion-exchange columns.

  • Contains three proteins for benchmarking anion exchange columns.
  • Designed for use with high-resolution IEX columns for monitoring performance.
  • Utilizes ion exchange chromatography with UV detection.
  • Solid formulation, supplied lyophilized in 20 mM Tris Buffer, pH 8–9.
  • Reconstitute with 1 mL of 100% Milli-Q Water.
  • Includes Horse Myoglobin (0.5 mg/mL), Conalbumin (1.25 mg/mL), and Soybean Trypsin Inhibitor (2 mg/mL).
  • Shelf life of 12 months; ships ambient.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.