Learn More
Waters Corp IEX Cation Test Standard 186006870
Supplier:  Waters Corp 186006870
Encompass_Preferred
A specialized mix of 3 Intact proteins; Ribonuclease A , Cytochrome C,and Chicken Lysozyme that is formulated to benchmark and test Cation Exchange Columns. This standard is a lyophilized powder packaged in a Waters Truview Max Recovery Vial. It is recommended that this standard be used on a regular basis for benchmarking column and system performance, similar to the QCRM product line.
Catalog No. NC1781049