Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp IEX-MS Start Up Kit, BioResolve SCX mAb Column, 3 μm, 2.1 x 100 mm

Catalog No. 502392598 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This IEX-MS start up kit features a 3 μm corrosion–resistant column (2.1 x 100 mm) for reproducible, high-resolution, charge-based separations of mAbs and other proteins in LC and LC-MS applications. The kit also includes a Humanized mAb Mass Check Standard and pH buffer concentrates.

  • Column type: BioResolve SCX mAb
  • Particle size: 3 μm
  • Column dimensions: 2.1 x 100 mm
  • Separation modes: UPLC, UHPLC, HPLC
  • Detection method compatibility: Optical, MS
  • Number of analysis: 50
  • Packages per kit: 1
  • System type: UPLC, HPLC
  • Units per package: 4 pk
  • Included components: BioResolve SCX mAb 3 μm corrosion–resistant column, Humanized mAb mass check standard, pH concentrate A (pH 5.0), pH concentrate B (pH 8.5)

Catalog No. 50-239-2598 Supplier Waters Corp Supplier No. 176004502
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This IEX-MS start up kit features a 3 μm corrosion–resistant column (2.1 x 100 mm) for reproducible, high-resolution, charge-based separations of mAbs and other proteins in LC and LC-MS applications. The kit also includes a Humanized mAb Mass Check Standard and pH buffer concentrates.

  • Column type: BioResolve SCX mAb
  • Particle size: 3 μm
  • Column dimensions: 2.1 x 100 mm
  • Separation modes: UPLC, UHPLC, HPLC
  • Detection method compatibility: Optical, MS
  • Number of analysis: 50
  • Packages per kit: 1
  • System type: UPLC, HPLC
  • Units per package: 4 pk
  • Included components: BioResolve SCX mAb 3 μm corrosion–resistant column, Humanized mAb mass check standard, pH concentrate A (pH 5.0), pH concentrate B (pH 8.5)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.