Learn More
Waters Corp IEX-MS Start Up Kit, BioResolve SCX mAb 3 μm, 2.1 x 100 mm Column
Catalog No. 502392826 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This IEX-MS Start Up Kit includes a corrosion–resistant column with polymer-based particles and sulfonic acid ligands for high-resolution, charge-based separations of monoclonal antibodies and other proteins in LC/LC-MS applications. It also provides a humanized mAb mass check standard and pH buffer concentrates.
  • Features a corrosion–resistant column for high-resolution separations.
  • Provides reproducible, charge-based separations for mAbs and proteins.
  • Compatible with LC and LC-MS applications using salt or pH gradient methods.
  • Includes a humanized mAb mass check standard.
  • Contains IonHance CX-MS pH Buffer Concentrates (pH 5.0 and pH 8.5) in MS certified LDPE containers.
  • Designed for 50 analyses; optimized for protein applications.
  • Supports UPLC, UHPLC, and HPLC system types with optical and MS detection.
Catalog No. 50-239-2826 Supplier Waters Corp Supplier No. 176004501
This IEX-MS Start Up Kit includes a corrosion–resistant column with polymer-based particles and sulfonic acid ligands for high-resolution, charge-based separations of monoclonal antibodies and other proteins in LC/LC-MS applications. It also provides a humanized mAb mass check standard and pH buffer concentrates.
  • Features a corrosion–resistant column for high-resolution separations.
  • Provides reproducible, charge-based separations for mAbs and proteins.
  • Compatible with LC and LC-MS applications using salt or pH gradient methods.
  • Includes a humanized mAb mass check standard.
  • Contains IonHance CX-MS pH Buffer Concentrates (pH 5.0 and pH 8.5) in MS certified LDPE containers.
  • Designed for 50 analyses; optimized for protein applications.
  • Supports UPLC, UHPLC, and HPLC system types with optical and MS detection.