Learn More
Medchemexpress LLC Ifebemtinib (hydrochloride) | 1227948-82-4 | 99.2% | 625.01 | 10 MM/1 ML
SDP
Catalog No. 5000437118
Encompass_Preferred
Ifebemtinib is an orally bioavailable inhibitor of the non-receptor, cytoplasmic tyrosine kinase protein tyrosine kinase 2 (FAK: PTK2) with potential antineoplastic activity. It targets and inhibits PTK2 in an adenosine triphosphate (ATP)-competitive manner, preventing PTK2-mediated downstream signaling and inhibiting migration, proliferation, invasion, and survival in PTK2-overexpressing tumor cells.
  • Orally bioavailable inhibitor of FAK1/PTK2
  • Targets and inhibits PTK2 in an ATP-competitive manner
  • Prevents PTK2-mediated downstream signaling
  • Inhibits migration, proliferation, invasion, and survival in PTK2-overexpressing tumor cells
  • Stable under recommended storage conditions
Catalog No. 50-004-37118 Supplier Medchemexpress LLC Supplier No. HY122844A10MM/1ML
Ifebemtinib is an orally bioavailable inhibitor of the non-receptor, cytoplasmic tyrosine kinase protein tyrosine kinase 2 (FAK: PTK2) with potential antineoplastic activity. It targets and inhibits PTK2 in an adenosine triphosphate (ATP)-competitive manner, preventing PTK2-mediated downstream signaling and inhibiting migration, proliferation, invasion, and survival in PTK2-overexpressing tumor cells.
  • Orally bioavailable inhibitor of FAK1/PTK2
  • Targets and inhibits PTK2 in an ATP-competitive manner
  • Prevents PTK2-mediated downstream signaling
  • Inhibits migration, proliferation, invasion, and survival in PTK2-overexpressing tumor cells
  • Stable under recommended storage conditions