Learn More
Medchemexpress LLC Ifebemtinib (hydrochloride) | 1227948-82-4 | 99.2% | 625.01 | 100 MG
SDP
Catalog No. 5000437120
Encompass_Preferred
Ifebemtinib hydrochloride is a potent and orally active FAK inhibitor. It is extensively researched for its role in inhibiting cancer progression.
  • Inhibits focal adhesion kinase (FAK) activity.
  • Demonstrates high potency with an IC50 of 1 nM for recombinant FAK.
  • Orally active, making it suitable for in vivo studies.
  • Potential for use in anticancer research.
  • Acts as an ATP-competitive inhibitor.
Catalog No. 50-004-37120 Supplier Medchemexpress LLC Supplier No. HY122844A100MG
Ifebemtinib hydrochloride is a potent and orally active FAK inhibitor. It is extensively researched for its role in inhibiting cancer progression.
  • Inhibits focal adhesion kinase (FAK) activity.
  • Demonstrates high potency with an IC50 of 1 nM for recombinant FAK.
  • Orally active, making it suitable for in vivo studies.
  • Potential for use in anticancer research.
  • Acts as an ATP-competitive inhibitor.