Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ifebemtinib hydrochloride | MFCD34676862 | 99.2% | 625.01 g/mol | C28H29ClF4N6O4 | 5 MG
SDP

Catalog No. 5000264537
Encompass_Preferred

Ifebemtinib hydrochloride is the hydrochloride salt of a selective focal adhesion kinase (FAK) inhibitor supplied for research use. It is provided as a white to off-white solid in small pack sizes for in vitro and early-stage studies, with specifications suitable for laboratory experiments.

  • High purity (99.22%).
  • Hydrochloride salt form for improved solubility in polar solvents.
  • White to off-white solid suitable for handling and formulation.
  • Available in small research pack sizes including 5 mg.
  • Molecular weight 625.01 g/mol; formula C28H29ClF4N6O4.

Catalog No. 50-002-64537 Supplier Medchemexpress LLC Supplier No. HY122844A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ifebemtinib hydrochloride is the hydrochloride salt of a selective focal adhesion kinase (FAK) inhibitor supplied for research use. It is provided as a white to off-white solid in small pack sizes for in vitro and early-stage studies, with specifications suitable for laboratory experiments.

  • High purity (99.22%).
  • Hydrochloride salt form for improved solubility in polar solvents.
  • White to off-white solid suitable for handling and formulation.
  • Available in small research pack sizes including 5 mg.
  • Molecular weight 625.01 g/mol; formula C28H29ClF4N6O4.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.