Learn More
Abcam Ifenprodil hemitartrate, GluN2B (formerly NR2B)-preferring NMDA antagonist, 50MG
Catalog No. 502707871
Encompass_Preferred
MW 801 Da, Purity >99%. Non-competitive NMDA receptor antagonist acting at the polyamine site. Displays GluN2B (formerly NR2B) subunit selectivity. IC₅₀ values are 0.15 μM at NR1/NR2B and >30 μM for NR1/NR2A, NR1/NR2C and NR1/NR2D receptors. Also available in simple stock solutions (ab146675) - add 1 ml of water to get an exact, ready-to-use concentration.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7871 Supplier Abcam Supplier No. AB12011150MG
MW 801 Da, Purity >99%. Non-competitive NMDA receptor antagonist acting at the polyamine site. Displays GluN2B (formerly NR2B) subunit selectivity. IC₅₀ values are 0.15 μM at NR1/NR2B and >30 μM for NR1/NR2A, NR1/NR2C and NR1/NR2D receptors. Also available in simple stock solutions (ab146675) - add 1 ml of water to get an exact, ready-to-use concentration.
The product is subject to the following: Abcam Restricted Use Statement