Learn More
Apexbio Technology LLC Ifenprodil Tartrate 23210-58-4 50mg
SDP
Catalog No. 5000092647
Encompass_Preferred
Ifenprodil Tartrate (CAS 23210-58-4) is a small molecule recognized as an atypical noncompetitive antagonist of the NMDA (N-methyl-D-aspartate) receptor It demonstrates high-affinity binding to a uniform population of NMDA receptors in neonatal rat forebrain tissue exhibiting an IC50 value of 0 3 M By selectively modulating NMDA receptor activity Ifenprodil Tartrate serves as a valuable tool in neuropharmacological research particularly for investigating glutamatergic neurotransmission excitotoxicity and the roles of NMDA receptor subtypes in neuronal development and pathology
Catalog No. 50-000-92647 Supplier Apexbio Technology LLC Supplier No. B162350
Ifenprodil Tartrate (CAS 23210-58-4) is a small molecule recognized as an atypical noncompetitive antagonist of the NMDA (N-methyl-D-aspartate) receptor It demonstrates high-affinity binding to a uniform population of NMDA receptors in neonatal rat forebrain tissue exhibiting an IC50 value of 0 3 M By selectively modulating NMDA receptor activity Ifenprodil Tartrate serves as a valuable tool in neuropharmacological research particularly for investigating glutamatergic neurotransmission excitotoxicity and the roles of NMDA receptor subtypes in neuronal development and pathology