Learn More
Medchemexpress LLC Ifenprodil (tartrate) | 23210-58-4 | 99.9% | 100 MG
SDP
Catalog No. 5000340288
Encompass_Preferred
Ifenprodil tartrate (NP-120) is a cerebral vasodilator and a noncompetitive NMDA receptor antagonist. It shows high affinity for NR1A/NR2B receptors and acts as an α1 adrenergic receptor antagonist, inhibiting GIRK (Kir3) to reduce inward currents. It exhibits reliable inhibitory effects against A/H1N1 strains and possesses neuroprotective, anticonvulsant, and antinociceptive properties.
  • Cerebral vasodilator
  • Noncompetitive NMDA receptor antagonist
  • High affinity for NR1A/NR2B receptors
  • Acts as an α1 adrenergic receptor antagonist
  • Inhibits GIRK (Kir3) activity
  • Exhibits inhibitory effects against A/H1N1 strains
  • Possesses neuroprotective, anticonvulsant, and antinociceptive properties
  • Useful for studying cerebrovascular diseases and peripheral arterial obliterative disease
Catalog No. 50-003-40288 Supplier Medchemexpress LLC Supplier No. HY12882A100MG
Ifenprodil tartrate (NP-120) is a cerebral vasodilator and a noncompetitive NMDA receptor antagonist. It shows high affinity for NR1A/NR2B receptors and acts as an α1 adrenergic receptor antagonist, inhibiting GIRK (Kir3) to reduce inward currents. It exhibits reliable inhibitory effects against A/H1N1 strains and possesses neuroprotective, anticonvulsant, and antinociceptive properties.
  • Cerebral vasodilator
  • Noncompetitive NMDA receptor antagonist
  • High affinity for NR1A/NR2B receptors
  • Acts as an α1 adrenergic receptor antagonist
  • Inhibits GIRK (Kir3) activity
  • Exhibits inhibitory effects against A/H1N1 strains
  • Possesses neuroprotective, anticonvulsant, and antinociceptive properties
  • Useful for studying cerebrovascular diseases and peripheral arterial obliterative disease