Learn More
Selleck Chemical LLC Ifenprodil Tartrate S4091-50mg
SDP
Catalog No. 503111261
Encompass_Preferred
Ifenprodil is an atypical noncompetitive antagonist at the NMDA receptor it interacts with high affinity at a homogeneous population of NMDA receptors in neonatal rat forebrain with IC50 of 0 3 M
Catalog No. 50-311-1261 Supplier Selleck Chemical LLC Supplier No. S409150MG
Ifenprodil is an atypical noncompetitive antagonist at the NMDA receptor it interacts with high affinity at a homogeneous population of NMDA receptors in neonatal rat forebrain with IC50 of 0 3 M