Learn More
Medchemexpress LLC HY-12882A 50mg Medchemexpress, Ifenprodil (tartrate) CAS:23210-58-4 Purity:>98%
SDP
Catalog No. 502081186
Encompass_Preferred
Medchemexpress, HY-12882A 50mg Ifenprodil (tartrate) CAS:23210-58-4 Ifenprodil tartrate is a typical noncompetitive NMDA receptor antagonist. Ifenprodil tartrate exerts high affinity at NR1A/NR2B receptors (IC50=0.34 μM) over 400-fold than at NR1A/NR2A receptors (IC50=146 μM). Ifenprodil tartrate inhibits GIRK (Kir3), reduces inward currents through the basal GIRK activity. Ifenprodil tartrate has the potential to be a cerebral vasodilator. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1186 Supplier Medchemexpress LLC Supplier No. HY12882A50MG
Add to Cart
Add to Cart
Medchemexpress, HY-12882A 50mg Ifenprodil (tartrate) CAS:23210-58-4 Ifenprodil tartrate is a typical noncompetitive NMDA receptor antagonist. Ifenprodil tartrate exerts high affinity at NR1A/NR2B receptors (IC50=0.34 μM) over 400-fold than at NR1A/NR2A receptors (IC50=146 μM). Ifenprodil tartrate inhibits GIRK (Kir3), reduces inward currents through the basal GIRK activity. Ifenprodil tartrate has the potential to be a cerebral vasodilator. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.