Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ProSci IFI16 (CT) Peptide
SDP

Catalog No. 16595425
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-595-425 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-595-425 Supplier ProSci Supplier No. 9641P0050
Only null left
Add to Cart
Add to Cart

Used for Blocking Activity

IFI16 (CT) Peptide is used for blocking the activity of IFI16 (CT) Antibody.

IFI16 Antibody: IFI16 inhibits DNA repair that potentiates type-I interferon-induced antitumor effects in triple negative breast cancer. Increased expression of IFI16 predicts adverse prognosis in multiple myeloma. IFI16 isoforms with cytoplasmic and nuclear locations play differential roles in recognizing invaded DNA viruses. IFI16 directly senses viral RNA and enhances RIG-I transcription and activation to restrict influenza virus infection. Nuclear sensor IFI16 inhibits the function of Hepatitis B virus covalently closed circular DNA by integrating innate immune activation and epigenetic suppression.

Specifications

Gene ID (Entrez) 3428
Species Human
Accession Number Q16666.3
Common Name interferon gamma inducible protein 16
Concentration 200 μg/mL
Content And Storage Store ULBP1 peptide at -20°C, stable for one year.
Format Liquid
Formulation PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin and 0.02% sodium azide
For Use With (Application) Blocking Assay
Gene Symbol IFI16
Quantity 0.05 mg
Sequence 16 amino acids near the carboxyl terminus of human IFI16.
Synonym IFI16 Antibody: Gamma-interferon-inducible protein 16, Ifi-16, Interferon-inducible myeloid differentiation transcriptional activator, IFNGIP1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.