Learn More
Aviva Systems Biology Corp IFI35 Antibody - N-terminal region (ARP46134_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46134_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IFI35. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human IFI35. Alternate names: FLJ21753, IFP35ApplicationWB. Species reactivity: Human
Catalog No. 50-014-53921