Learn More
Aviva Systems Biology Corp IFI44 Antibody - middle region (ARP46154_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46154_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IFI44. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human IFI44. Alternate names: MTAP44, p44ApplicationWB. Species reactivity: Human
Catalog No. 50-014-53920