Learn More
Aviva Systems Biology Corp IFIH1 Antibody - middle region (ARP36486_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36486_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IFIH1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human IFIH1. Alternate names: Hlcd, IDDM19, MDA-5, MDA5, MGC133047ApplicationIHC, WB. Species reactivity: Cow, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-56171