Learn More
Aviva Systems Biology Corp IFIT2 Antibody - N-terminal region (ARP41950_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41950_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IFIT2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human IFIT2. Alternate names: G10P2, GARG-39, IFI-54, IFI54, ISG-54K, ISG54, cig42, IFIT-2, IFI-54K, ISG-54 KApplicationWB. Species reactivity: Human
Catalog No. 50-014-58060